Anti-S100 alpha Mouse Monoclonal Antibody [clone: ABT-S100A1]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (ABT-S100A1) antibody to S100A1 for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: S100 alpha
Clonality: Monoclonal
Clone: ABT-S100A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77081-320
- 77075-458
- Antigen Symbol:
- S100 alpha
- S100 alpha
- Antigen Name:
- Bpb
- Bpb
- Antigen Synonyms:
- S-100 protein subunit alpha|S100 Calcium Binding Protein A1|S100 Alpha Chain|Protein S100-A1|NEF|S-100 protein alpha chain|S100|S100 Beta Chain|S100 alpha
- S-100 protein subunit alpha|S100 Calcium Binding Protein A1|S100 Alpha Chain|Protein S100-A1|NEF|S-100 protein alpha chain|S100|S100 Beta Chain|S100 alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-S100A1
- ABT-S100A1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P23297
- UniprotID# P23297
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human S100A1
- Synthetic peptide derived from human S100A1
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage Buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC
- IHC
Specifications
Related Information
Frequently Bought Together