Anti-FHIT Rabbit Polyclonal Antibody
Catalog # 77071-120
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AP3A hydrolase
- Antigen Symbol:FHIT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P49789
- Antigen Synonyms:AP3Aase|Bis 5' adenosyl triphosphatase|AP3A hydrolase fragile site 3p14.2|Bis(5' adenosyl) triphosphatase|Dinucleosidetriphosphatase|FRA3B|FHIT|FRA 3B|Diadenosine 5' 5''' P1 P3 triphosphate hydrolase
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:16 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human FHIT (amino acids 81-130)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77071-120
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to FHIT for WB, IHC, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: FHIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human