Anti-USP24 Rabbit Polyclonal Antibody
Catalog # 77076-114
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Deubiquitinating enzyme 24
- Antigen Symbol:USP24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UPU5
- Antigen Synonyms:Ubiquitin carboxyl terminal hydrolase 24|Ubiquitin carboxyl-terminal hydrolase 24|Ubiquitin thioesterase 24|Ubiquitin specific processing protease 24|KIAA1057|Ubiquitin specific protease 24|Ubiquitin-specific-processing protease 24|Ubiquitin thiolesterase 24|Ubiquitin specific peptidase 24
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:211 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human USP24 (amino acids 21-70)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-114
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to USP24 for IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: USP24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse