Anti-Inactive ubiquitin carboxyl-terminal hydrolase 50 Rabbit Polyclonal Antibody
Catalog # 77080-262
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Inactive ubiquitin carboxyl-terminal hydrolase 50
- Antigen Symbol:Inactive ubiquitin carboxyl-terminal hydrolase 50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q70EL3
- Antigen Synonyms:Ubiquitin specific protease 50|Inactive ubiquitin specific peptidase 50|USP50
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:38 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human USP50 (amino acids 251-300)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77080-262
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to USP50 for IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: Inactive ubiquitin carboxyl-terminal hydrolase 50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse