Anti-ELOVL4 Rabbit Polyclonal Antibody
77061-918
- Antibody Type:Primary
- Antigen Name:3-keto acyl-CoA synthase ELOVL4
- Antigen Symbol:ELOVL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9GZR5
- Antigen Synonyms:ELOV L4|Elongation of very long chain fatty acids like 4|Cancer/testis antigen 118|CT118|ELOV4_HUMAN|Elongation of very long chain fatty acids protein 4|Elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast) like 4|ELOVL 4|ADMD
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ELOVL4 (amino acids 41-90)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77061-918
- Packaging:Plastic vial
Rabbit polyclonal antibody to ELOVL4 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: ELOVL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse