Anti-POLR3G Rabbit Polyclonal Antibody
77078-724
- Antibody Type:Primary
- Antigen Name:DNA directed RNA polymerase III 32 kDa polypeptide
- Antigen Symbol:POLR3G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O15318
- Antigen Synonyms:DNA-directed RNA polymerase III subunit RPC7|RPC7_HUMAN|RNA polymerase III subunit C7|DNA-directed RNA polymerase III subunit G|Polymerase (RNA) III (DNA directed) polypeptide G (32kD)|RPC32|POLR3G|RNA polymerase III 32 kDa subunit
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:25 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human POLR3G (amino acids 151-200)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77078-724
- Packaging:Plastic vial
Rabbit polyclonal antibody to POLR3G for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: POLR3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse