Anti-DNA Ligase IV Rabbit Polyclonal Antibody
77075-952
- Antibody Type:Primary
- Antigen Name:DNA joinase
- Antigen Symbol:DNA Ligase IV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P49917
- Antigen Synonyms:LIG 4|LIG4S|DNA ligase 4|DNA repair enzyme|DNA Ligase IV|Ligase IV DNA ATP dependent|DNLI4_HUMAN|DNA ligase IV ATP dependent|LIG4
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:103 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human DNA Ligase 4 (amino acids 616-665)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77075-952
- Packaging:Plastic vial
Rabbit polyclonal antibody to DNA Ligase 4 for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: DNA Ligase IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human