Order Entry
United States
Orders LinkContactUsLinkComponent
 

Anti-ZNF608 Rabbit Polyclonal Antibody

100 µg, Plastic vial
Catalog # 77063-332
Supplier:   Antibodies.com
 :  A98619-100
Anti-ZNF608 Rabbit Polyclonal Antibody
Catalog # 77063-332
Supplier:  Antibodies.com
Anti-ZNF608 Rabbit Polyclonal Antibody
Anti-ZNF608 Rabbit Polyclonal Antibody

100 µg, Plastic vial

Price
$514.78
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Rabbit polyclonal antibody to ZNF608 for IHC and ELISA with samples derived from Human and Mouse.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100 ELISA: 1:5000

 : This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse

Specifications
  • Size:
    100 µg
  • Packaging:
    Plastic vial
  • Clonality:
    Polyclonal
  • Molecular Weight:
    162 kDa
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Form:
    Liquid
  • Storage Buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Tested Applications:
    IHC
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cat. No.:
    77063-332
  • Reactivity:
    Human|Mouse
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Antibody Type:
    Primary
  • Gene ID:
    UniprotID# Q9ULD9
  • Immunogen:
    Synthetic peptide derived from human ZNF608 (amino acids 1231-1280)
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Storage Temperature:
    Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.