Anti-Angiopoietin like 7 Rabbit Polyclonal Antibody
77063-024
- Antibody Type:Primary
- Antigen Name:Angiopoietin like 7
- Antigen Symbol:Angiopoietin like 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O43827
- Antigen Synonyms:Angiopoietin-related protein 7|Angiopoietin like factor (CDT6)|AngX|ANGL7_HUMAN|Angiopoietin-like factor|Angiopoietin like factor|ANGPTL7|Angiopoietin related protein 7|Angiopoietin-like protein 7
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ANGPTL7 (amino acids 297-346)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-024
- Packaging:Plastic vial
Rabbit polyclonal antibody to ANGPTL7 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Angiopoietin like 7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse