Anti-BRCC45 Rabbit Polyclonal Antibody
Catalog # 77075-216
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:6030405P19Rik
- Antigen Symbol:BRCC45
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NXR7
- Antigen Synonyms:brain and reproductive organ expressed protein|BRCA1-A complex subunit BRE|Brain and reproductive organ expressed (TNFRSF1A modulator)|BRCA1/BRCA2 containing complex subunit 4|B830038C02Rik|AI429776|BRCA1 A complex subunit BRE|BRCA1/BRCA2 containing complex subunit 45|Brain and reproductive organ-expressed protein
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:46 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BRE (amino acids 325-374)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77075-216
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to BRE for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: BRCC45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat