Anti-IL1RAPL2 Rabbit Polyclonal Antibody
Catalog # 77067-002
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:IL 1 RAPL 2
- Antigen Symbol:IL1RAPL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NP60
- Antigen Synonyms:IL-1R9|IL 1R 9|IL-1RAPL-2|IL-1R-9|IL 1 receptor accessory protein like 2|IL-1 receptor accessory protein-like 2|IL 1R9|IL 1RAPL 2|IL-1-RAPL-2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:78 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human IL1RAPL2 (amino acids 141-190)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-002
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to IL1RAPL2 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: IL1RAPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse