Anti-Bone marrow stromal cell antigen 1 Rabbit Polyclonal Antibody
Catalog # 77063-986
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ADP ribosyl cyclase 2
- Antigen Symbol:Bone marrow stromal cell antigen 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q10588
- Antigen Synonyms:ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2|BST1|Bone marrow stromal antigen 1|cADPr hydrolase 2|Bone marrow stromal cell antigen 1|BST-1|BST 1|BST1_HUMAN|ADP-ribosyl cyclase 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BST1 (amino acids 71-120)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-986
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to BST1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: Bone marrow stromal cell antigen 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat