Anti-GRK5 Rabbit Polyclonal Antibody
Catalog # 77061-004
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:FLJ39780
- Antigen Symbol:GRK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P34947
- Antigen Synonyms:G protein coupled receptor kinase 5|G protein-coupled receptor kinase 5|GRK-pan|G protein coupled receptor kinase GRK5|Pan-GRK|GRK5_HUMAN|FP2025|GPRK5|G protein-coupled receptor kinase GRK5
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:67 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GRK5 (amino acids 351-400)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77061-004
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GRK5 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: GRK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat