Anti-5HT7 Receptor Rabbit Polyclonal Antibody
77060-914
- Antibody Type:Primary
- Antigen Name:5 HT 7
- Antigen Symbol:5HT7 Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P34969
- Antigen Synonyms:5HT7|5-HT7|5 hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase coupled)|5 HT X|5-HT-7|5 hydroxytryptamine receptor 7|5 hydroxytryptamine (serotonin) receptor 7|5-HT-X|5-hydroxytryptamine receptor 7
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:53 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human HTR7 (amino acids 391-440)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77060-914
- Packaging:Plastic vial
Rabbit polyclonal antibody to HTR7 for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: 5HT7 Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat