Anti-PKA regulatory subunit I beta Rabbit Polyclonal Antibody
Catalog # 77060-838
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:cAMP dependent protein kinase type I beta regulatory subunit
- Antigen Symbol:PKA regulatory subunit I beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P31321
- Antigen Synonyms:cAMP-dependent protein kinase type I-beta regulatory subunit|Prkar1b|KAP1_HUMAN|PRKAR1|PRKAR1B protein|PRKAR 1|PKARI beta|PRKAR 1B|PKA regulatory subunit I beta
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:43 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human KAP1 (amino acids 106-155)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77060-838
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to KAP1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: PKA regulatory subunit I beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat