Anti-SLC27A5 Rabbit Polyclonal Antibody
77059-842
- Antibody Type:Primary
- Antigen Name:ACSB
- Antigen Symbol:SLC27A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y2P5
- Antigen Synonyms:BA-CoA ligase|Bile acid-CoA ligase|BAL|BA CoA ligase|ACSVL6|Bile acyl CoA synthetase|BACS|Bile acyl-CoA synthetase|Bile acid CoA ligase
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC27A5 (amino acids 481-530)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77059-842
- Packaging:Plastic vial
Rabbit polyclonal antibody to SLC27A5 for WB, IHC, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: SLC27A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human