Anti-ADI1 Rabbit Polyclonal Antibody
77059-730
- Antibody Type:Primary
- Antigen Name:1 2 dihydroxy 3 keto 5 methylthiopentene dioxygenase
- Antigen Symbol:ADI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9BV57
- Antigen Synonyms:ADI 1|Acireductone dioxygenase (Ni(2+)-requiring)|ADI1|2-dihydroxy-3-keto-5-methylthiopentene dioxygenase|Acireductone dioxygenase (Ni(2+) requiring)|Aci reductone dioxygenase|Acireductone dioxygenase 1|Acireductone dioxygenase (Fe(2+) requiring)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:21 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human ADI1 (amino acids 71-120)
- Tested Applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77059-730
- Packaging:Plastic vial
Rabbit polyclonal antibody to ADI1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC-P: 1:50 to 1:300, ELISA: 1:10000
Type: Primary
Antigen: ADI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat