Anti-Apolipoprotein O like Rabbit Polyclonal Antibody
77059-386
- Antibody Type:Primary
- Antigen Name:AAIR8193
- Antigen Symbol:Apolipoprotein O like
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6UXV4
- Antigen Synonyms:MGC129748|Apolipoprotein O-like|CXorf33|APOOL|Family with sequence similarity 121A|Protein FAM121A|Apolipoprotein O like|APOOL_HUMAN|FAM121A
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:29 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human APOOL (amino acids 219-268)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77059-386
- Packaging:Plastic vial
Rabbit polyclonal antibody to APOOL for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: Apolipoprotein O like
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat