Anti-BMP8a Rabbit Polyclonal Antibody
Catalog # 77059-294
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BMP-8A
- Antigen Symbol:BMP8a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z5Y6
- Antigen Synonyms:FLJ14351|Bone morphogenetic protein 8A|BMP8A_HUMAN|FLJ45264|OP 2|BMP8a|Osteogenic protein 2|BMP8
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:44 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human BMP8A (amino acids 241-290)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Cat. No.:77059-294
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to BMP8A for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: BMP8a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat