Anti-Myozenin 2 Rabbit Polyclonal Antibody
77058-134
- Antibody Type:Primary
- Antigen Name:C4orf5
- Antigen Symbol:Myozenin 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NPC6
- Antigen Synonyms:Calsarcin1|CS 1|CMH16|Calsarcin-1|Calcineurin binding protein calsarcin 1|CS1|Muscle specific protein|FATZ related protein 2|FATZ-related protein 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:29 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human MYOZ2 (amino acids 21-70)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77058-134
- Packaging:Plastic vial
Rabbit polyclonal antibody to MYOZ2 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 , ELISA: 1:10000
Type: Primary
Antigen: Myozenin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat