Anti-TEAD1 + TEAD2 + TEAD3 + TEAD4 Rabbit Polyclonal Antibody
Catalog # 77057-232
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DTEF 1
- Antigen Symbol:TEAD1 + TEAD2 + TEAD3 + TEAD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15562
- Antigen Synonyms:TEA domain family member 1|TEA domain family member 4|TEA domain family member 3|TCF13|Protein GT-IIC|NTEF 1|TEA domain family member 2|TCF 13|SV40 transcriptional enhancer factor
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:49 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TEAD2 (amino acids 71-120)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77057-232
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TEAD2 for IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: TEAD1 + TEAD2 + TEAD3 + TEAD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse