Anti-CARD10 Rabbit Polyclonal Antibody
Catalog # 77056-926
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Bcl10 binding protein and activator of NFKB
- Antigen Symbol:CARD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9BWT7
- Antigen Synonyms:Carma 3|CARD containing MAGUK 3 protein|CARD 10|CARD10|CARD-containing MAGUK protein 3|CARMA3|BIMP1|CARD-containing MAGUK 3|CAR10_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:115 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CARD10 (amino acids 481-530)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77056-926
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CARD10 for WB, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: CARD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse