Anti-CCT6A Rabbit Polyclonal Antibody
Catalog # 77056-730
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Acute morphine dependence related protein 2
- Antigen Symbol:CCT6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P40227
- Antigen Synonyms:CCT-zeta-1|CCT6A|CCT-zeta|Chaperonin containing T complex subunit 6|CCT6|CCT zeta 1|CCT zeta|Amino acid transport defect complementing|CCTZ
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:58 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CCT6A (amino acids 171-220)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77056-730
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CCT6A for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: CCT6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat