Anti-IQGAP3 Rabbit Polyclonal Antibody
77055-962
- Antibody Type:Primary
- Antigen Name:IQ motif containing GTPase activating protein 3
- Antigen Symbol:IQGAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q86VI3
- Antigen Synonyms:Ras GTPase activating like protein IQGAP3|MGC10170|IQGAP3|MGC1947|IQGA3_HUMAN|OTTHUMP00000031855|IQGAP 3|MGC10831|OTTHUMP00000031854
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:184 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human IQGAP3 (amino acids 90-139)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77055-962
- Packaging:Plastic vial
Rabbit polyclonal antibody to IQGAP3 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: IQGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat