Anti-SLC9A7 Rabbit Polyclonal Antibody
77055-466
- Antibody Type:Primary
- Antigen Name:Na(+)/H(+) exchanger 7
- Antigen Symbol:SLC9A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q96T83
- Antigen Synonyms:SLC9A6|SLC9A7|NHE-7|Nonselective sodium potassium/proton exchanger|Sodium/hydrogen exchanger 7|Solute carrier family 9 member 7|NHE7|SL9A7_HUMAN|Solute carrier family 9 (sodium/hydrogen exchanger), member 7
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:80 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC9A7 (amino acids 531-580)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77055-466
- Packaging:Plastic vial
Rabbit polyclonal antibody to SLC9A7 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: SLC9A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat