Anti-RABEP1 Rabbit Polyclonal Antibody
Catalog # 77055-042
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Neurocrescin
- Antigen Symbol:RABEP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15276
- Antigen Synonyms:RABEP1|RAB5EP|RABPT5A|Renal carcinoma antigen NY REN 17|Rabaptin 5|Rabaptin 4|Rab GTPase binding effector protein 1|RABPT5|Rabaptin 5alpha
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:99 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RABEP1 (amino acids 549-598)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77055-042
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to RABEP1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: RABEP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat