Anti-EPAC1 delta DEP Rabbit Polyclonal Antibody
Catalog # 77054-742
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:bcm910
- Antigen Symbol:EPAC1 delta DEP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O95398
- Antigen Synonyms:CGEF1|EPAC|EPAC1|cAMP GEFI|EPAC1 delta DEP|Epac 1|cAMP-GEFI|cAMP-regulated guanine nucleotide exchange factor I|cAMP regulated guanine nucleotide exchange factor I
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:99 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RAPGEF3 (amino acids 712-761)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77054-742
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to RAPGEF3 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: EPAC1 delta DEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat