Anti-Cytochrome P450 2D6 Rabbit Polyclonal Antibody
77054-534
- Antibody Type:Primary
- Antigen Name:CPD6
- Antigen Symbol:Cytochrome P450 2D6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10635
- Antigen Synonyms:CYP2D7BP|CYP2DL1|CYP2D7AP|Cytochrome P450 2D6|CYP2D8P2|CYP2D7P2|CYP2D6|CYPIID6|CYP2D
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:55 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Cytochrome P450 2D6 (amino acids 228-277)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77054-534
- Packaging:Plastic vial
Rabbit polyclonal antibody to Cytochrome P450 2D6 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Cytochrome P450 2D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human