Anti-nNOS Rabbit Polyclonal Antibody
77054-054
- Antibody Type:Primary
- Antigen Name:2310005C01Rik
- Antigen Symbol:nNOS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P29475
- Antigen Synonyms:N-NOS|Constitutive NOS|NC-NOS|IHPS 1|neuronal Nitric Oxide Synthase|EC 1.14.13.39|BNOS|IHPS1|Neuronal NOS
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:160 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human nNOS (amino acids 818-867)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77054-054
- Packaging:Plastic vial
Rabbit polyclonal antibody to nNOS for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: nNOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat