Anti-LARG Rabbit Polyclonal Antibody
Catalog # 77053-930
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ARHGC_HUMAN
- Antigen Symbol:LARG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NZN5
- Antigen Synonyms:Rho guanine nucleotide exchange factor (GEF) 12|Rho guanine nucleotide exchange factor 12|Rho guanine exchange factor (GEF) 12|PRO2792|Leukemia-associated RhoGEF|KIAA0382|LARG|leukemia-associated rho guanine nucleotide exchange factor|ARHGEF12
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:173 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ARHGEF12 (amino acids 449-498)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77053-930
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ARHGEF12 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: LARG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat