Anti-Calcium channel L type DHPR alpha 2 subunit Rabbit Polyclonal Antibody
Catalog # 77053-902
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CA2D1_HUMAN
- Antigen Symbol:Calcium channel L type DHPR alpha 2 subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P54289
- Antigen Synonyms:CACNL2A|CCHL2A|Cacna2d1|CACN A2|CACNA2|Calcium channel L type DHPR alpha 2 subunit|Calcium channel voltage dependent alpha 2/delta subunit 1|Dihydropyridine receptor alpha 2 subunit|Calcium channel L type alpha 2 polypeptide
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:123 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Cacna2d1 (amino acids 90-139)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77053-902
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Cacna2d1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Calcium channel L type DHPR alpha 2 subunit
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat