Order Entry
United States
ContactUsLinkComponent
 
EGFR (phospho Thr678) Cell Based ELISA Kit
Catalog # 77213-732
Supplier:  Avantor
CAS Number:  
undefined
EGFR (phospho Thr678) Cell Based ELISA Kit
Catalog # 77213-732
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
    Rat
    Mouse
  • Target protein:
    EGFR
  • Description:
    EGFR (phospho Thr678) cell based ELISA kit
  • Size:
    2×96 tests
  • Sample Type:
    Adherent cells and suspension cells
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    >5000 cells
  • Storage Temperature:
    4 °C
  • Regulatory Status:
    RUO
  • Cat. No.:
    77213-732

Specifications

About this item

This EGFR (phospho Thr678) Cell Based ELISA kit allows for the detection of EGFR (phospho Thr678) and the effects that certain stimulation conditions have on EGFR (phospho Thr678) expression in different cell lines. Qualitative determination of EGFR (phospho Thr678) concentration is achieved by an indirect ELISA format. In essence, the EGFR (phospho Thr678) is captured by Anti-EGFR (phospho Thr678) Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this EGFR (phospho Thr678) Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH Antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted. 3) Anti-EGFR Antibody is provided for normalization purposes. The absorbance values obtained for the non-phosphorylated target can be used to normalize the absorbance values for the phosphorylated target.