Human Betacellulin ELISA Kit
Catalog # 77213-368
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Betacellulin
- Description:Human betacellulin ELISA kit
- Size:96 tests
- Sample Type:Plasma, serum and lysates
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:8 - 1000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77213-368
Specifications
About this item
This Human Betacellulin ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human Betacellulin in plasma, serum, and lysates.