Order Entry
United States
ContactUsLinkComponent
Mouse VEGFB ELISA Kit
Mouse VEGFB ELISA Kit
Catalog # 77201-204
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse VEGFB ELISA Kit
Catalog # 77201-204
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A310541-96
CAS Number:  

Specifications

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    30 - 480 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    1.61 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77201-204

Specifications

About this item

Mouse VEGFB ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse VEGFB in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 30 to 480 ng/L
  • Sensitivity: 1.61 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse VEGFB ELISA kit (A310541) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse VEGFB in An antibody specific for VEGFB has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the VEGFB present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-VEGFB antibody, which also binds the VEGFB present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-VEGFB antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-VEGFB antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of VEGFB captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of VEGFB in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.