Order Entry
United States
ContactUsLinkComponent
Mouse VE Cadherin ELISA Kit
Mouse VE Cadherin ELISA Kit
Catalog # 77173-972
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse VE Cadherin ELISA Kit
Catalog # 77173-972
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313522-96
CAS Number:  

Specifications

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1 - 16 ng/mL
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.024 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77173-972

Specifications

About this item

Mouse VE Cadherin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse VE Cadherin in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1 to 16 ng/ml
  • Sensitivity: 0.024 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse VE Cadherin ELISA kit (A313522) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse VE Cadherin in An antibody specific for VE Cadherin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the VE Cadherin present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-VE Cadherin antibody, which also binds the VE Cadherin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-VE Cadherin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-VE Cadherin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of VE Cadherin captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of VE Cadherin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.