Human TRAIL ELISA Kit
Catalog # 77207-070
Supplier: Avantor
CAS Number:
Specifications
- Detection Method:Chemiluminescent (Luminometer)
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:8 - 500 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77207-070