Order Entry
United States
ContactUsLinkComponent
Human TARC/CCL17 ELISA Kit
Human TARC/CCL17 ELISA Kit
  77173-832
 :  
undefined
Human TARC/CCL17 ELISA Kit
  77173-832
 :  A313452-96
 :  

 

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    150 - 2400 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    5.51 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77173-832

 

 

Human TARC/CCL17 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TARC/CCL17 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 5.51 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human TARC/CCL17 ELISA kit (A313452) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TARC/CCL17 in An antibody specific for TARC/CCL17 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TARC/CCL17 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-TARC/CCL17 antibody, which also binds the TARC/CCL17 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-TARC/CCL17 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-TARC/CCL17 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of TARC/CCL17 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of TARC/CCL17 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.