Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse Rspo3 ELISA Kit

Mouse Rspo3 ELISA Kit

Mouse Rspo3 ELISA kit, 96 tests
Catalog # 77211-646
Supplier:   Antibodies.com
Supplier #:  A311202-96
Mouse Rspo3 ELISA Kit
Catalog # 77211-646
Supplier:  Antibodies.com
CAS Number:  
Mouse Rspo3 ELISA Kit
Mouse Rspo3 ELISA Kit

Mouse Rspo3 ELISA kit, 96 tests

CAS Number:  
Price
$591.17
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mouse Rspo3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Rspo3 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 20 to 320 ng/L
  • Sensitivity: 0.97 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Rspo3 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Rspo3 in serum, plasma or other biological fluids. An antibody specific for Rspo3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Rspo3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Rspo3 antibody, which also binds the Rspo3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Rspo3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Rspo3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Rspo3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Rspo3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Mouse Rspo3 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    20 - 320 ng/L
  • Sample Volume:
    40 μl
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    77211-646
  • Regulatory Status:
    RUO
  • Assay duration:
    One step
  • Sensitivity:
    0.97 ng/L
  • Format:
    Pre-coated
  • Target protein:
    Rspo3
  • Sample Type:
    Serum, plasma or other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.