Order Entry
United States
ContactUsLinkComponent
Human Retinoic acid Receptor beta ELISA Kit
Human Retinoic acid Receptor beta ELISA Kit
Catalog # 77172-988
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Human Retinoic acid Receptor beta ELISA Kit
Catalog # 77172-988
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313026-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Retinoic acid Receptor beta
  • Description:
    Human Retinoic acid receptor beta ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    40 - 640 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    2.16 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77172-988

Specifications

About this item

Human Retinoic acid Receptor beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Retinoic acid Receptor beta in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 40 to 640 ng/L
  • Sensitivity: 2.16 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Retinoic acid Receptor beta ELISA kit (A313026) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Retinoic acid Receptor beta in serum, plasma or other biological fluids. An antibody specific for Retinoic acid Receptor beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Retinoic acid Receptor beta present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Retinoic acid Receptor beta antibody, which also binds the Retinoic acid Receptor beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Retinoic acid Receptor beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Retinoic acid Receptor beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Retinoic acid Receptor beta captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Retinoic acid Receptor beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.