Order Entry
United States
ContactUsLinkComponent
REQU Cell Based ELISA Kit
REQU Cell Based ELISA Kit
Catalog # 77203-114
Supplier:  Avantor
CAS Number:  
undefined
REQU Cell Based ELISA Kit
Catalog # 77203-114
Supplier:  Avantor
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
    Mouse
  • Target protein:
    REQU
  • Description:
    REQU cell based ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Adherent cells and suspension cells.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    >5000 Cells
  • Storage Temperature:
    4 °C
  • Regulatory Status:
    RUO
  • Cat. No.:
    77203-114

Specifications

About this item

This REQU Cell Based ELISA kit allows for the detection of REQU and the effects that certain stimulation conditions have on REQU expression in different cell lines. Qualitative determination of REQU concentration is achieved by an indirect ELISA format. In essence, the REQU is captured by Anti-REQU Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this REQU Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted.