Order Entry
United States
ContactUsLinkComponent
Human Placental Lactogen ELISA Kit
Human Placental Lactogen ELISA Kit
Catalog # 77209-916
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Placental Lactogen ELISA Kit
Catalog # 77209-916
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A311331-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Placental Lactogen
  • Description:
    Human Placental Lactogen ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.5 - 24 µg/mL
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.052 µg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77209-916

Specifications

About this item

Human Placental Lactogen ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Placental Lactogen in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1.5 to 24 µg/ml
  • Sensitivity: 0.052 µg/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Placental Lactogen ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Placental Lactogen in serum, plasma or other biological fluids. An antibody specific for Placental Lactogen has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Placental Lactogen present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Placental Lactogen antibody, which also binds the Placental Lactogen present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Placental Lactogen antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Placental Lactogen antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Placental Lactogen captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Placental Lactogen in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.