Human P-Cadherin ELISA Kit
Catalog # 77201-812
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:P-Cadherin
- Description:Human P-Cadherin ELISA kit
- Size:96 tests
- Detection Method:Chemiluminescent (Luminometer)
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15 - 1000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77201-812