Human MMP-7 ELISA Kit
Catalog # 77204-040
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:MMP-7
- Description:Human MMP-7 ELISA kit
- Size:96 tests
- Detection Method:Chemiluminescent (Luminometer)
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:8 - 500 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77204-040