ContactUsLinkComponent
TIF-IA (phospho Ser649) Cell Based ELISA Kit
TIF-IA (phospho Ser649) Cell Based ELISA Kit
Catalog # 77201-274
Supplier:  Avantor
CAS Number:  
undefined
TIF-IA (phospho Ser649) Cell Based ELISA Kit
Catalog # 77201-274
Supplier:  Avantor
CAS Number:  
Specifications
  • Cat. No.:
    77201-274
Specifications
About this item
This TIF-IA (phospho Ser649) Cell Based ELISA kit allows for the detection of TIF-IA (phospho Ser649) and the effects that certain stimulation conditions have on TIF-IA (phospho Ser649) expression in different cell lines. Qualitative determination of TIF-IA (phospho Ser649) concentration is achieved by an indirect ELISA format. In essence, the TIF-IA (phospho Ser649) is captured by Anti-TIF-IA (phospho Ser649) Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this TIF-IA (phospho Ser649) Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH Antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted. 3) Anti-TIF-IA Antibody is provided for normalization purposes. The absorbance values obtained for the non-phosphorylated target can be used to normalize the absorbance values for the phosphorylated target.