ContactUsLinkComponent
Caspase 3 (phospho Ser150) Cell Based ELISA Kit
Caspase 3 (phospho Ser150) Cell Based ELISA Kit
Catalog # 77204-726
Supplier:  Avantor
CAS Number:  
undefined
Caspase 3 (phospho Ser150) Cell Based ELISA Kit
Catalog # 77204-726
Supplier:  Avantor
CAS Number:  
Specifications
  • Cat. No.:
    77204-726
Specifications
About this item
This Caspase 3 (phospho Ser150) Cell Based ELISA kit allows for the detection of Caspase 3 (phospho Ser150) and the effects that certain stimulation conditions have on Caspase 3 (phospho Ser150) expression in different cell lines. Qualitative determination of Caspase 3 (phospho Ser150) concentration is achieved by an indirect ELISA format. In essence, the Caspase 3 (phospho Ser150) is captured by Anti-Caspase 3 (phospho Ser150) Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this Caspase 3 (phospho Ser150) Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH Antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted. 3) Anti-Caspase 3 Antibody is provided for normalization purposes. The absorbance values obtained for the non-phosphorylated target can be used to normalize the absorbance values for the phosphorylated target.