- Sample Type:Adherent cells and suspension cells
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:>5000 cells
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77204-960
This FGFR1 (phospho Tyr766) Cell Based ELISA kit allows for the detection of FGFR1 (phospho Tyr766) and the effects that certain stimulation conditions have on FGFR1 (phospho Tyr766) expression in different cell lines. Qualitative determination of FGFR1 (phospho Tyr766) concentration is achieved by an indirect ELISA format. In essence, the FGFR1 (phospho Tyr766) is captured by Anti-FGFR1 (phospho Tyr766) Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this FGFR1 (phospho Tyr766) Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH Antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted. 3) Anti-FGFR1 Antibody is provided for normalization purposes. The absorbance values obtained for the non-phosphorylated target can be used to normalize the absorbance values for the phosphorylated target.