Order Entry
United States
Orders LinkContactUsLinkComponent
Human HGF ELISA Kit

Human HGF ELISA Kit

Catalog # 77211-576
Human HGF ELISA Kit
Catalog # 77211-576
Supplier:  
CAS Number:  
Human HGF ELISA Kit
Human HGF ELISA Kit
CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This Human HGF ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human HGF in plasma, serum, and lysates.

Specifications

Specifications

  • Cat. No.:
    77211-576
  • Regulatory Status:
    RUO
  • Sample Type:
    Plasma, serum and lysates
  • Detection Range:
    125 - 8000 pg/ml
  • Shelf Life:
    Store for 6 months at 4 °C
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Storage Temperature:
    4 °C

Specifications