Human MIA-2 ELISA Kit
Catalog # 77203-436
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:MIA-2
- Description:Human MIA-2 ELISA kit
- Size:96 tests
- Sample Type:Plasma, serum and lysates
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:16 - 2000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77203-436
Specifications
About this item
This Human MIA-2 ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human MIA-2 in plasma, serum, and lysates.