ContactUsLinkComponent
Human Pro-Cathepsin B ELISA Kit
Human Pro-Cathepsin B ELISA Kit
Catalog # 77215-998
Supplier:  Avantor
CAS Number:  
undefined
Human Pro-Cathepsin B ELISA Kit
Catalog # 77215-998
Supplier:  Avantor
CAS Number:  
Specifications
  • Cat. No.:
    77215-998
Specifications
About this item
This Human Pro-Cathepsin B ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human Pro-Cathepsin B in plasma, serum, and lysates.