ContactUsLinkComponent
Human SCFR ELISA Kit
  77209-700
 :  Avantor
 :  
undefined
Human SCFR ELISA Kit
  77209-700
 :  Avantor
 :  
  • Cat. No.:
    77209-700
This Human SCFR ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human SCFR in plasma, serum, and lysates.